Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- How the kit works:
- In this experiment, the identity of a metal carbonate is determined by gravimetric analysis, a technique that measures mass to quantify an analyte. By comparing the masses of two compounds containing the analyte, the formula weight and identity of the unknown metal carbonate can be determined
- Kit Includes:
- 1 x 10 g Calcium Chloride, Anhydrous
- 1 x 5 g Silver Nitrate
Specifications
Specifications
| Grade Level | High School, Higher Ed |
| Grade | 9 to 12 |
| Class Size | 15 Lab Groups |
| Product Type | AP Chemistry Lab Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?