Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- How the kit works:
- In this experiment, benzoic acid is prepared by the Grignard method. The Grignard reagent is first carboxylated by reaction with dry ice to form the target carboxylic acid. After the reaction is complete, hot water is added to remove any remaining solvent, and acid is added to dissolve the magnesium salts and liberate the benzoic acid, which is then collected upon cooling
- Kit Includes:
- 1 x 5 g Magnesium Metal Turnings
- 2 x 25 mL Tetrahydrofuran
- 1 x 20 mL Bromobenzene
- 1 x 25 mL Hydrochloric Acid, 36°C
- 1 x 10 g Decolorizing Carbon
Specifications
Specifications
| Grade Level | High School, Higher Ed |
| Grade | 9 to 12 |
| Class Size | 15 Lab Groups |
| Product Type | AP Chemistry Lab Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?