Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Structure and Concentration In Acid and Base Titrations
GREENER_CHOICESCI_ED

Catalog No. S07323 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S07323 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.
Catalog No. S07323 Supplier Innovating Science™ Supplier No. IS8114
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.

Includes

0.2M Hydrochloric Acid Part A (4 x 500mL), 0.1M Sulfuric Acid Part B (4 x 500mL), 0.05M Nitric Acid Part C (2 x 1L), 0.1M Calcium Hydroxide (4 x 500mL), 0.2M Ammonium Hydroxide Part E (4 x 500mL), 0.1N Acetic Acid Part F (4 x 500mL), 0.1N Sodium Hydroxide (4 x 500mL), teacher's guide, student study guide copymasters

Requires

Beakers (50, 150 and 250mL), graduated cylinders (10 and 100mL), pH meter, stir bar, test tubes (16 x 150mm), wash bottle, buret (50mL), magnetic stirrer, pH test paper, support stand and buret clamp, test tube rack, distilled or deionized water

Conduct a series of acid–base titrations and determine the concentrations of two unknowns.

Students begin by finding the pH of a solution with pH paper. Next they create a procedure to collect quantitative titration data using a buret and pH meter. Using titration data that they collected, they can determine the concentration of each unknown.

  • Meets Big Idea 6, Investigation 14, Primary Learning Objective 6.13
  • Contains materials for 15 groups of students

Specifications

Certifications/Compliance Meets big idea 6, investigation 14, primary learning objective 6.13
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To determine the concentrations of two unknowns
Includes 4 x 500mL 0.2M Hydrochloric Acid, Part A, 4 x 500mL 0.1M Sulfuric Acid, Part B, 2 x 1 L 0.05M Nitric Acid, Part C, 4 x 500mL Calcium Hydroxide 0.1M, 4 x 500mL Ammonium Hydroxide 0.2M, Part E, 4 x 500mL 0.1N Acetic Acid, Part F, 4 x 500mL 0.1N Sodium Hydroxide, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 18.5 x 14.5 x 14.5 in.
Class Size 15 Lab Groups
Product Type Structure and Concentration In Acid and Base Titrations Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.