Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: What's In That Bottle?
GREENER_CHOICESCI_ED

Catalog No. S07315 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S07315 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, and n-Hexane, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S07315 Supplier Innovating Science™ Supplier No. IS8106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, and n-Hexane, which are known to the State of California to cause cancer and reproductive harm.

Includes

10g Ammonium Chloride, 10g Magnesium Oxide, 10g Benzoic Acid, 10g Aluminum Metal Powder 200 mesh, 10g Paraffin Wax, 10g Sodium Acetate, 10g Sodium Carbonate Anhy Soda Ash, 10g Copper Metal, 50 mesh, 99% Min, 10g Cupric Sulfate Pentahy XL's, 10g Sodium Chloride,10g Calcium Carbonate, 10g Potassium Nitrate, 10g Salicylic Acid, 10g Calcium Metal Granular 8-Mesh, 10g Zinc Metal, 10g Cupric Sulfate, 10g Sucrose Granular Sugar, 10g Sodium Bicarbonate, 10g coated Magnesium granules, 10g Urea Crystals, 30mL Universal Indicator, 30mL Sodium Hydroxide 0.1N, 30mL N-Hexanes, 30mL Deionized water, 30mL Hydrochloric Acid 0.1N, 30mL Ethyl Alcohol 95% Denatured, 30mL Phenolphthalein 1% Ipa, 32 Capillary tubes

Demonstrates physical and chemical properties of unknown chemicals through various lab tests.

Students will identify the 4 different kinds of bonds that exist in chemicals: ionic, polar covalent, nonpolar covalent and metallic. Students review the properties of each solid using various tests.

  • Meets Big Idea 1, Investigation 6 and Primary Learning Objective 2.22
  • Contains materials for 15 groups of students and teacher's guide and student study guide copymasters

Order Info

This package conforms to 49 CFR 173.4 for domestic highway or rail transport only.

Specifications

Certifications/Compliance Meets big idea 2, investigation 6, primary learning objective 2.22
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Demonstrates physical and chemical properties of unknown chemicals through various lab tests
Includes 10g Ammonium Chloride, 10g Magnesium Oxide, 10g Benzoic Acid, 10g Aluminum Metal, 10g Paraffin wax, 10g Sodium Acetate, 10g Sodium Carbonate, 10g Copper Metal, 10g Cupric Sulfate, Pentahydrate, 10g Sodium Chloride, 30mL universal indicator, 30mL 0.1M Sodium Hydroxide, 30mL Hexane, 30mL deionized water, LED conductivity tester, 10g Calcium Carbonate, 10g Potassium Nitrate, 10g Salicylic Acid, 10g Calcium Metal, 10g Zinc Metal, 10g Cupric Sulfate, anhydrous, 10g Sucrose, 10g Sodium Bicarbonate, 10g Magnesium metal, 10g Urea, 30mL 0.1M Hydrochloric Acid, 30mL 95% Ethanol, 30mL 1% Phenolphthalein, 32 Melting point tubes, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 12.75 x 9.5 x 6.75 in.
Class Size 15 Lab Groups
Product Type What'S In That Bottle Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.