Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Liquid Column Chromatography

Catalog No. S001291 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size Grade
S001291 15 Lab Groups 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Glass wool fibers (inhalable and biopersistent), which are known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.
Catalog No. S001291 Supplier Innovating Science™ Supplier No. IS8031
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Glass wool fibers (inhalable and biopersistent), which are known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.

This hands-on laboratory activity kit is ideal for mastering column separation and equilibrium principles.

  • How the kit works:
    • In column chromatography, the mixture to be analyzed is applied to the top of the column, and the eluent is passed through by gravity or air pressure. As equilibrium is established between the adsorbed solutes and the flowing solvent, the mixture components move at different rates due to their varying interactions with the stationary and mobile phases, resulting in separation. The individual components are then collected as they elute from the bottom of the column
  • Kit Includes:
    • 4 X 25 mL Isopropyl Alcohol
    • 1 X 100 g Silica gel 60
    • 2 X 15 mL Dye Mixture
    • 15 Pipettes
    • Glass Wool

Specifications

Grade Level High School, Higher Ed
Grade 9 to 12
Class Size 15 Lab Groups
Product Type AP Chemistry Lab Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.