Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Thermochromism

Catalog No. S001295 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size Grade
S001295 15 Lab Groups 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Mercury/mercury compounds, which are known to the State of California to cause reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. S001295 Supplier Innovating Science™ Supplier No. IS8019
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Mercury/mercury compounds, which are known to the State of California to cause reproductive harm. For more information go to www.P65Warnings.ca.gov.

Explore the science of color change in this laboratory activity. Students synthesize metal complexes to observe how heat alters crystal structure and light absorption. This reversible, visual experiment is a captivating way to study transition metals and thermodynamics.

  • How the kit works:
    • The way a crystalline material absorbs or reflects light depends on its crystal structure, which can change when heat is applied. In this experiment, two compounds formed by reacting metal salts with the [HgI₄]²⁻ ion are heated on a hot plate, causing a color change. Upon cooling, the compounds return to their original color, demonstrating a reversible change in crystal structure
  • Kit Includes:
    • 3 x 25 g Mercury (II) Chloride
    • 3 x 25 g Potassium Iodide
    • 1 x 15 g Silver Nitrate
    • 2 x 25 g Copper (II) Chloride, Anhydrous

Specifications

Grade Level High School, Higher Ed
Grade 9 to 12
Class Size 15 Lab Groups
Product Type AP Chemistry Lab Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.