Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- How the kit works:
- Commercial food colors may contain more components than are visible to the eye. In this experiment, students use thin layer chromatography to analyze four different food colors and investigate their composition
- By comparing results obtained with three different solvents, students can draw conclusions about the solubility of each dye. The kit contains sufficient materials for 15 groups and includes a Teacher's Manual and Student Study Guide copy-masters
- Kit Includes:
- 1 Food coloring sample set containing:
- 1 x 0.5 mL Yellow
- 1 x 0.5 mL Red
- 1 x 0.5 mL Blue
- 1 x 0.5 mL Green
- 5 x 30 mL Chromatography Solvent no.1 (Deionized water)
- 5 x 30 mL Chromatography Solvent no.2 (50°C Ethanol)
- 5 x 30 mL Chromatography Solvent no.3 (95°C Ethanol)
- 15 TLC (Thin Layer Chromatography) Sheets
- 4 Capillary Pipettes
Specifications
Specifications
| Grade Level | High School, Higher Ed |
| Grade | 9 to 12 |
| Class Size | 15 Lab Groups |
| Product Type | AP Chemistry Lab Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?