Learn More
Waters Corp AP Column Tubing Adapter, 2/pk
Supplier:  Waters Corp WAT021951
Encompass_Preferred
Waters Advanced Purification (AP) columns feature non-metallic construction and adjustable bed height with easy to use coarse and fine adjustments. The AP Glass columns are available in a variety of dimensions. Glass Tubes, Column Jackets, Plastic Shields and other replacement parts are sold separately.
Catalog No. 50-785-135