Learn More
Seracare Life Sciences Inc AP ELISA KIT ANTI-HUMAN
Supplier:  Seracare Life Sciences Inc 51100012
Encompass_Preferred
Highly sensitive reagents optimized for antigen detection in picogram quantities. Pretested wash and blocking solutions ensure low background. Can be used for both direct and indirect sandwich assays. Protein Detector ELISA Kits offer a convenient starting point for the development of microwell ELISA protocols for antigen and antibody detection. These kits provide pretested components and suggested protocols to facilitate the development of ELISA procedures. KPL's protein Detector ELISA Kit, AP Systems are provided with BluePhos Phosphatase Substrate which offers greater sensitivty, wider dyanmic range, and lower background than pNPP. BluePhos is also very convenient to use as a 2-Component systerm; simply mix and use.
Catalog No. 50-675-31