Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Environmental Science: Landfills and Decomposition
GREENER_CHOICESCI_ED

Catalog No. S000828 Shop All Aldon Products
Change view
Click to view available options
Product Type:
Landfills and Decomposition
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S000828 Landfills and Decomposition
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000828 Supplier Innovating Science™ Supplier No. IS9900
Add to Cart
Add to Cart

Simulate a sanitary landfill and an open-air dumpyard to explore the advantages of a landfill while observing decomposition in both aerobic and anaerobic environments.

  • Simulate a sanitary landfill and an open-air dumpyard to explore the advantages of a landfill while observing decomposition in both aerobic and anaerobic environments.
  • This kit contains enough materials for 8 groups of students and comes with a Teacher's manual and Student Study Guide Copymaster.
  • The activity covers AP Environmental Science primary learning objectives STB-3.K and STB-3.L.
  • It will take a total of 2 to 3 hours, with over a 5-day wait time to complete this activity (Day 1: 1.5 to 2 hours, Days 2 through 5+: 10 to 15 minutes each).

The kit contains:

  • Soil, 3000 cm3
  • 10 Plastic Cups, 9 oz.
  • 8 Plastic Jars with Lids
  • Cotton Fabric 3 x 6 in.
  • 8 Starch Packing Peanuts
  • 32 Paper Squares
  • 4 Straws
  • 16 Aluminum Squares

Needed but not supplied:

  • Graduated Cylinders, 25 mL
  • Markers (for labeling)
  • Metric Rulers
  • Scissors or Razor
  • Leaves, Bread, Fruit, etc., are optional (for additional organic waste sources)

Specifications

Grade Level High School
Grade 9 to 12
For Use With (Application) Education
Class Size 8 Lab Groups
Product Type Landfills and Decomposition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.