Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Environmental Science: Ocean Acidification
GREENER_CHOICESCI_ED

Catalog No. S000826 Shop All Aldon Products
Change view
Click to view available options
Product Type:
Ocean Acidification
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S000826 Ocean Acidification
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000826 Supplier Innovating Science™ Supplier No. IS9904
Add to Cart
Add to Cart

Simulate ocean acidification by introducing carbon dioxide into simulated ocean water while measuring the pH over time.

Changes to the carbon cycle and climate due to human activities have had detrimental effects on the environment. One example of this is known as ocean acidification, which is the changing of the ocean's chemistry due to increased carbon dioxide in the atmosphere and ocean. In this activity, simulate ocean acidification by introducing carbon dioxide into simulated ocean water while measuring the pH over time.

  • This kit contains enough materials for 8 groups of students and comes with a Teacher's Manual and a Student Study Guide Copymaster.
  • The activity covers AP Environmental Science primary learning objective STB-4.H.
  • It will take approximately 1.5 to 2 hours to complete this activity in a single day.

Kit contains:

  • Simulated Ocean Water QuickPrep, for 1 Gallon
  • Sodium Carbonate, 40 g
  • 2 Baking Soda, 100 g
  • 2 Vinegar, 500 mL
  • 8 Pipettes
  • 8 Tygon Tubing, 40 cm
  • 8 Rubber #7 Stoppers, 2-Hole
  • 8 Air Stones
  • 8 Spoons

Needed but not supplied:

  • Distilled Water
  • Tap Water
  • 8 Erlenmeyer Flasks,500 mL
  • 8 Beakers, 125 mL
  • 8 Graduated Cylinders, 100 mL
  • 8 pH Meters
  • 8 Stopwatches
  • Laboratory Balance
  • Masking Tape
  • Stir Bars, Optional
  • Stir Plates, Optional

Specifications

Grade Level High School
Grade 9 to 12
Class Size 8 Lab Groups
Product Type Ocean Acidification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.