Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Environmental Science: Water Quality
SCI_ED

Catalog No. S000825 Shop All Aldon Products
Change view
Click to view available options
Product Type:
Water Quality
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S000825 Water Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid, Cadmium and cadmium compounds, and Methanol, which are known to the State of California to cause cancer or reproductive harm.
Catalog No. S000825 Supplier Innovating Science™ Supplier No. IS9903
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid, Cadmium and cadmium compounds, and Methanol, which are known to the State of California to cause cancer or reproductive harm.

Test a local water body for a variety of parameters to determine overall water quality and learn about different indicators and sources of pollution.

Test a local water body for various parameters to determine overall water quality and learn about different indicators and sources of pollution.

  • This kit contains enough materials for 8 groups of students and comes with a Teacher's manual and Student Study Guide Copymaster.
  • The activity covers AP Environmental Science primary learning objectives STB-3.B and STB-3.F.
  • It will take a total of about 3 hours with a 5-day wait time to complete this activity (Day 1: 190 minutes, Day 2/3: 5 minutes, Day 4: 5 minutes, Day 5: 45 minutes).

Kit Contains:

  • 8 Collection Bottles, 500 mL
  • 16 DO/BOD Sample Bottles, 4 oz.
  • 8 Coliform Test Powder Vials; Griess Reagent, 15 mL
  • Cadmium Metal Powder, 0.5 g
  • 2 Manganese Chloride Reagent, 25 mL
  • Alkaline Iodide Reagent, 40 mL
  • Sulfuric Acid, 50%, 40 mL
  • Starch Indicator Solution, 25 mL
  • Sodium Thiosulfate Titrant, 500 mL
  • 8 Phosphate Powder Pillows
  • Wide Range pH Test Strips, pk. of 100
  • Toothpicks, pk. of 100
  • 8 Secchi Sticks

Needed but not supplied:

  • Markers
  • Aluminum Foil
  • Distilled Water
  • Graduated Cylinder, 25 mL
  • 125 mL Erlenmeyer Flasks
  • Funnels
  • Burettes, 50 mL
  • Burette Clamp/Stands
  • TDS/Conductivity Meter, Optional
  • pH Meter, Optional

Specifications

Grade Level High School
Grade 9 to 12
For Use With (Application) Education
Class Size 8 Lab Groups
Product Type Water Quality
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.