Learn More
MSC Rite-Kem Cleaner and Degreaser, 55 gal, Water-Based, Drum
Catalog No. 502839172
Encompass
This cleaner and degreaser is formulated with emulsifiers, dispersants, and penetrants designed to break down grease and grime. It effectively separates the bond between surfaces and soil, allowing for easy rinsing, and is suitable for cleaning plastic, rubber, and metal.
  • Formulated with emulsifiers, dispersants, and penetrants
  • Breaks the bond between surface and soil for easy rinsing
  • Effectively cleans plastic, rubber, and metal
  • Comes in a 55 gallon drum
  • Water-based and dilutable concentrate
Catalog No. 50-283-9172 Supplier MSC Supplier No. 48175871
This cleaner and degreaser is formulated with emulsifiers, dispersants, and penetrants designed to break down grease and grime. It effectively separates the bond between surfaces and soil, allowing for easy rinsing, and is suitable for cleaning plastic, rubber, and metal.
  • Formulated with emulsifiers, dispersants, and penetrants
  • Breaks the bond between surface and soil for easy rinsing
  • Effectively cleans plastic, rubber, and metal
  • Comes in a 55 gallon drum
  • Water-based and dilutable concentrate