Learn More
Medchemexpress LLC HY-15858 1mg , AP-III-a4 CAS:1177827-73-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY158581MG
Encompass_Preferred
HY-15858 1mg AP-III-a4 CAS: 1177827-73-4 AP-III-a4 (ENOblock) is a nonsubstrate analogue enolase inhibitor with an IC50 of 0.576 uM. AP-III-a4 can be used for the research of cancer and diabetic. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0956