Learn More
Selleck Chemical LLC AP-III-a4
SDP
Catalog No. 5000001361
Encompass_Preferred
AP-III-a4 (ENOblock) is the first nonsubstrate analogue inhibitor of enolase with IC50 of 0 576 M
Catalog No. 50-000-01361 Supplier Selleck Chemical LLC Supplier No. S744310MM/1ML
AP-III-a4 (ENOblock) is the first nonsubstrate analogue inhibitor of enolase with IC50 of 0 576 M