Learn More
Selleck Chemical LLC AP-III-a4 (ENOblock) 2mg 2070014-95-6
SDP
Supplier:  Selleck Chemical LLC S74432MG
Encompass_Preferred
AP-III-a4 (ENOblock) is the first nonsubstrate analogue inhibitor of enolase with IC50 of 0.576 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4779