Learn More
Medchemexpress LLC Ap-III-a4 hydrochloride | 2070014-95-6 | 99.9% | 631.18 g/mol | C31H44ClFN8O3 | 10 MG
SDP
Catalog No. 5000288624
Encompass_Preferred
AP-III-a4 hydrochloride is a small-molecule, nonsubstrate analogue inhibitor of enolase supplied as the hydrochloride salt for research use. It is reported to bind and inhibit enolase (IC50 0.576 μM) and has been used in studies of cancer cell viability, migration, invasion, apoptosis, and glucose metabolism.
  • Nonsubstrate analogue enolase inhibitor (IC50 0.576 μM).
  • Used in cancer and diabetes research, including HCT116 cell assays.
  • Shown to inhibit cell migration and invasion and to induce apoptosis.
  • Increases glucose uptake and inhibits PEPCK expression in cell studies.
  • Supplied as a hydrochloride solid suitable for laboratory assays, commonly available in small quantities such as 10 MG.
  • Analytical documentation typically available (HPLC, LCMS, certificate of analysis).
Catalog No. 50-002-88624 Supplier Medchemexpress LLC Supplier No. HY15858A10MG
AP-III-a4 hydrochloride is a small-molecule, nonsubstrate analogue inhibitor of enolase supplied as the hydrochloride salt for research use. It is reported to bind and inhibit enolase (IC50 0.576 μM) and has been used in studies of cancer cell viability, migration, invasion, apoptosis, and glucose metabolism.
  • Nonsubstrate analogue enolase inhibitor (IC50 0.576 μM).
  • Used in cancer and diabetes research, including HCT116 cell assays.
  • Shown to inhibit cell migration and invasion and to induce apoptosis.
  • Increases glucose uptake and inhibits PEPCK expression in cell studies.
  • Supplied as a hydrochloride solid suitable for laboratory assays, commonly available in small quantities such as 10 MG.
  • Analytical documentation typically available (HPLC, LCMS, certificate of analysis).