Learn More
Anton Paar Viscosity Standard Anton Paar N415 (H) 100 mL - 1100/330/32 mPa.s
Catalog No. NC0845013
Encompass_Preferred
Viscosity Standard AP N415c for adjustments and check measurements. Exact viscosity and density values are submitted with an UKAS accredited certificate, according to ISO/IEC 17025
Catalog No. NC0845013 Supplier Anton Paar Supplier No. 100048
Viscosity Standard AP N415c for adjustments and check measurements. Exact viscosity and density values are submitted with an UKAS accredited certificate, according to ISO/IEC 17025