Learn More
Leica Biosystems AP Purple Chromogen - Ready-to-use alkaline phosphatase chromogen for IHC, 50 tests
Catalog No. NC3916371
Encompass
Leica Biosystems AP Purple Chromogen (DC9459) is a ready-to-use alkaline phosphatase chromogenic substrate formulated for immunohistochemistry (IHC) and in situ hybridization (ISH) research applications. It produces a distinct purple precipitate optimized for multiplex staining workflows and is supplied in a kit sized for approximately 50 tests. Intended for use on automated research stainers; research use only.
  • Ready-to-use alkaline phosphatase chromogen for IHC and ISH.
  • Produces a purple precipitate suitable for multiplex staining.
  • Formulated for automated research stainers.
  • Supplied quantity for approximately 50 tests.
  • Intended for research use only.
  • Compatible with combination use alongside other AP or peroxidase substrates for multiplexing.
Catalog No. NC3916371 Supplier Leica Biosystems Supplier No. DC9459
Leica Biosystems AP Purple Chromogen (DC9459) is a ready-to-use alkaline phosphatase chromogenic substrate formulated for immunohistochemistry (IHC) and in situ hybridization (ISH) research applications. It produces a distinct purple precipitate optimized for multiplex staining workflows and is supplied in a kit sized for approximately 50 tests. Intended for use on automated research stainers; research use only.
  • Ready-to-use alkaline phosphatase chromogen for IHC and ISH.
  • Produces a purple precipitate suitable for multiplex staining.
  • Formulated for automated research stainers.
  • Supplied quantity for approximately 50 tests.
  • Intended for research use only.
  • Compatible with combination use alongside other AP or peroxidase substrates for multiplexing.