Learn More
Seracare Life Sciences Inc AP STABILIZER
Catalog No. 5067520
Encompass_Preferred
APStabilizer is effective at stabilizing phosphatase conjugates in diluted form for extended periods of time with minimum effect on conjugate activity. Stability studies conducted have indicated the APStabilizer retains greater stability of diluted AP conjugates than 50% glycerol, BSA or other commercially available products when stored at 37 C, room temperature or 4 C. Conjugates may be diluted to a use dilution concentration as low as 1 ug/mL and be stable for at least 6 months when stored at 4 C. Product in ready-to-use liquid form.
This item is not returnable. View return policy
Catalog No. 50-675-20 Supplier Seracare Life Sciences Inc Supplier No. 52900007
APStabilizer is effective at stabilizing phosphatase conjugates in diluted form for extended periods of time with minimum effect on conjugate activity. Stability studies conducted have indicated the APStabilizer retains greater stability of diluted AP conjugates than 50% glycerol, BSA or other commercially available products when stored at 37 C, room temperature or 4 C. Conjugates may be diluted to a use dilution concentration as low as 1 ug/mL and be stable for at least 6 months when stored at 4 C. Product in ready-to-use liquid form.