Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP Stop QT

Catalog No. QVT0517
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QVT0517 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QVT0517 Supplier Invitrogen™ Supplier No. QVT0517
Add to Cart
Add to Cart

AP Stop QT, 10 mL, is an aqueous buffered solution intended for the inactivation of residual LP6-AP activity after the Fast Blue Substrate development.

AP Stop QT, 10 mL, is an aqueous buffered solution intended for the inactivation of residual LP6-AP activity after the Fast Blue Substrate development. It is an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C
Product Type AP Stop QT Solution
Quantity 10 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.