Learn More
Surmodics Inc AP Yellow One Component Microwell Substrate (pNPP) with Stabilizer Pellets, 100mL
SDP
Catalog No. NC9444916
Encompass_Preferred
A one-component PNPP substrate that provides a soluble yellow product upon reaction with alkaline phosphatase in ELISA applications.The formulation consists of p-nitrophenylphosphate (p-NPP) solution with stabilizing pellets to for long term stability. The product is stable for 3 years from the date of manufacture.
Catalog No. NC9444916 Supplier Surmodics Inc Supplier No. PNPS010001
A one-component PNPP substrate that provides a soluble yellow product upon reaction with alkaline phosphatase in ELISA applications.The formulation consists of p-nitrophenylphosphate (p-NPP) solution with stabilizing pellets to for long term stability. The product is stable for 3 years from the date of manufacture.