Learn More
ABclonal Technology Phospho-Rac1/2/3-S71 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP00861000UL
Encompass_Preferred
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-10211