Learn More
ABclonal Technology Phospho-AKT1-S473 Rabbit pAb
SDP
Catalog No. 5000113246
Encompass_Preferred
This gene encodes a member of the AKT serine-threonine kinase family, involved in regulating cell proliferation, survival, metabolism, and angiogenesis. AKT proteins are activated by phosphorylation via phosphoinositide 3-kinase (PI3K) and play key roles in various signaling pathways, including those involving receptor tyrosine kinases and G-protein coupled receptors. AKT activation requires phosphorylation of threonine 308 and serine 473. The PI3K/AKT pathway is crucial for tumor cell survival by inhibiting apoptosis and activating survival factors. AKT also interacts with the mTOR pathway, affecting translation initiation and is often dysregulated in cancers. Mutations in this gene are linked to various cancers, Proteus syndrome, Cowden syndrome, and other growth disorders. Multiple transcript variants exist due to alternative splicing.
Catalog No. 50-001-13246 Supplier ABclonal Technology Supplier No. AP0098500UL
This gene encodes a member of the AKT serine-threonine kinase family, involved in regulating cell proliferation, survival, metabolism, and angiogenesis. AKT proteins are activated by phosphorylation via phosphoinositide 3-kinase (PI3K) and play key roles in various signaling pathways, including those involving receptor tyrosine kinases and G-protein coupled receptors. AKT activation requires phosphorylation of threonine 308 and serine 473. The PI3K/AKT pathway is crucial for tumor cell survival by inhibiting apoptosis and activating survival factors. AKT also interacts with the mTOR pathway, affecting translation initiation and is often dysregulated in cancers. Mutations in this gene are linked to various cancers, Proteus syndrome, Cowden syndrome, and other growth disorders. Multiple transcript variants exist due to alternative splicing.