Learn More
ABclonal Technology Phospho-GATA1-S310 Rabbit pAb
SDP
Catalog No. 5000110825
Encompass_Preferred
This gene encodes a protein which belongs to the GATA family of transcription factors. The protein plays an important role in erythroid development by regulating the switch of fetal hemoglobin to adult hemoglobin. Mutations in this gene have been associated with X-linked dyserythropoietic anemia and thrombocytopenia.
Catalog No. 50-001-10825 Supplier ABclonal Technology Supplier No. AP03541000UL
This gene encodes a protein which belongs to the GATA family of transcription factors. The protein plays an important role in erythroid development by regulating the switch of fetal hemoglobin to adult hemoglobin. Mutations in this gene have been associated with X-linked dyserythropoietic anemia and thrombocytopenia.