Learn More
ABclonal Technology Phospho-MARCKS-S162 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP04031000UL
Encompass_Preferred
The protein encoded by this gene is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.
Catalog No. 50-001-10822