Learn More
ABclonal Technology Phospho-α-Synuclein-S129 Rabbit pAb
SDP
Catalog No. 5000110318
Encompass_Preferred
Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. Synucleins are abundantly expressed in the brain and alpha- and beta-synuclein inhibit phospholipase D2 selectively. SNCA may serve to integrate presynaptic signaling and membrane trafficking. Defects in SNCA have been implicated in the pathogenesis of Parkinson disease. SNCA peptides are a major component of amyloid plaques in the brains of patients with Alzheimers disease. Alternatively spliced transcripts encoding different isoforms have been identified for this gene.
Catalog No. 50-001-10318 Supplier ABclonal Technology Supplier No. AP04501000UL
Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. Synucleins are abundantly expressed in the brain and alpha- and beta-synuclein inhibit phospholipase D2 selectively. SNCA may serve to integrate presynaptic signaling and membrane trafficking. Defects in SNCA have been implicated in the pathogenesis of Parkinson disease. SNCA peptides are a major component of amyloid plaques in the brains of patients with Alzheimers disease. Alternatively spliced transcripts encoding different isoforms have been identified for this gene.