Learn More
ABclonal Technology Phospho-Bcl-2-S70 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP05751000UL
Encompass_Preferred
This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-10923