Learn More
ABclonal Technology Phospho-GSK3α-S21 Rabbit pAb
SDP
Catalog No. 5000110939
Encompass_Preferred
This gene encodes a multifunctional Ser/Thr protein kinase that is implicated in the control of several regulatory proteins including glycogen synthase, and transcription factors, such as JUN. It also plays a role in the WNT and PI3K signaling pathways, as well as regulates the production of beta-amyloid peptides associated with Alzheimers disease.
Catalog No. 50-001-10939 Supplier ABclonal Technology Supplier No. AP05821000UL
This gene encodes a multifunctional Ser/Thr protein kinase that is implicated in the control of several regulatory proteins including glycogen synthase, and transcription factors, such as JUN. It also plays a role in the WNT and PI3K signaling pathways, as well as regulates the production of beta-amyloid peptides associated with Alzheimers disease.