Learn More
ABclonal Technology Phospho-STK11-S334 Rabbit pAb
SDP
Catalog No. 5000109553
Encompass_Preferred
The protein encoded by this gene is a serine/threonine kinase that regulates cell polarity and energy metabolism and functions as a tumor suppressor. Mutations in this gene have been associated with the autosomal dominant Peutz-Jeghers syndrome, as well as with skin, pancreatic, and testicular cancers.
Catalog No. 50-001-09553 Supplier ABclonal Technology Supplier No. AP0601500UL
The protein encoded by this gene is a serine/threonine kinase that regulates cell polarity and energy metabolism and functions as a tumor suppressor. Mutations in this gene have been associated with the autosomal dominant Peutz-Jeghers syndrome, as well as with skin, pancreatic, and testicular cancers.