Learn More
ABclonal Technology Phospho-ATR-S428 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP06761000UL
Encompass_Preferred
The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.
Catalog No. 50-001-11453