Learn More
ABclonal Technology Phospho-eIF4E-S209 Rabbit pAb
SDP
Catalog No. 5000113492
Encompass_Preferred
The protein encoded by this gene is a component of the eukaryotic translation initiation factor 4F complex, which recognizes the 7-methylguanosine cap structure at the 5 end of messenger RNAs. The encoded protein aids in translation initiation by recruiting ribosomes to the 5-cap structure. Association of this protein with the 4F complex is the rate-limiting step in translation initiation. This gene acts as a proto-oncogene, and its expression and activation is associated with transformation and tumorigenesis. Several pseudogenes of this gene are found on other chromosomes. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-13492 Supplier ABclonal Technology Supplier No. AP0747500UL
The protein encoded by this gene is a component of the eukaryotic translation initiation factor 4F complex, which recognizes the 7-methylguanosine cap structure at the 5 end of messenger RNAs. The encoded protein aids in translation initiation by recruiting ribosomes to the 5-cap structure. Association of this protein with the 4F complex is the rate-limiting step in translation initiation. This gene acts as a proto-oncogene, and its expression and activation is associated with transformation and tumorigenesis. Several pseudogenes of this gene are found on other chromosomes. Alternative splicing results in multiple transcript variants.