Learn More
ABclonal Technology Phospho-p90Rsk/RSK1/RPS6KA1-S363 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP0767500UL
Encompass_Preferred
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Catalog No. 50-001-10602