Learn More
ABclonal Technology Phospho-PEA15-S104 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP07881000UL
Encompass_Preferred
This gene encodes a death effector domain-containing protein that functions as a negative regulator of apoptosis. The encoded protein is an endogenous substrate for protein kinase C. This protein is also overexpressed in type 2 diabetes mellitus, where it may contribute to insulin resistance in glucose uptake. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-11289