Learn More
ABclonal Technology Phospho-AMPKα1-S485 Rabbit pAb
SDP
Catalog No. 5000110220
Encompass_Preferred
The protein encoded by this gene belongs to the ser/thr protein kinase family. It is the catalytic subunit of the 5-prime-AMP-activated protein kinase (AMPK). AMPK is a cellular energy sensor conserved in all eukaryotic cells. The kinase activity of AMPK is activated by the stimuli that increase the cellular AMP/ATP ratio. AMPK regulates the activities of a number of key metabolic enzymes through phosphorylation. It protects cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Catalog No. 50-001-10220 Supplier ABclonal Technology Supplier No. AP08711000UL
The protein encoded by this gene belongs to the ser/thr protein kinase family. It is the catalytic subunit of the 5-prime-AMP-activated protein kinase (AMPK). AMPK is a cellular energy sensor conserved in all eukaryotic cells. The kinase activity of AMPK is activated by the stimuli that increase the cellular AMP/ATP ratio. AMPK regulates the activities of a number of key metabolic enzymes through phosphorylation. It protects cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways. Alternatively spliced transcript variants encoding distinct isoforms have been observed.