Learn More
ABclonal Technology Phospho-GYS1-S641 Rabbit pAb
SDP
Catalog No. 5000109808
Encompass_Preferred
The protein encoded by this gene catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-09808 Supplier ABclonal Technology Supplier No. AP08771000UL
The protein encoded by this gene catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.