Learn More
ABclonal Technology Phospho-CREB1-S129 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP09031000UL
Encompass_Preferred
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds as a homodimer to the cAMP-responsive element, an octameric palindrome. The protein is phosphorylated by several protein kinases, and induces transcription of genes in response to hormonal stimulation of the cAMP pathway. Alternate splicing of this gene results in several transcript variants encoding different isoforms.
Catalog No. 50-001-12315