Learn More
ABclonal Technology Phospho-Smad2-S465/S467 Rabbit pAb
SDP
Catalog No. 5000109803
Encompass_Preferred
This gene encodes a protein in the SMAD family, which transduces signals and regulates transcription in various pathways. It mediates TGF-beta signaling, affecting cell proliferation, apoptosis, and differentiation. The protein interacts with the SMAD anchor for receptor activation (SARA) and is recruited to TGF-beta receptors. Phosphorylation by TGF-beta receptors causes dissociation from SARA and association with SMAD4, allowing translocation to the nucleus. There, it binds to promoters and forms a transcription repressor complex. The protein can also be phosphorylated by activin type 1 receptor kinase to mediate activin signaling. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-09803 Supplier ABclonal Technology Supplier No. AP09251000UL
This gene encodes a protein in the SMAD family, which transduces signals and regulates transcription in various pathways. It mediates TGF-beta signaling, affecting cell proliferation, apoptosis, and differentiation. The protein interacts with the SMAD anchor for receptor activation (SARA) and is recruited to TGF-beta receptors. Phosphorylation by TGF-beta receptors causes dissociation from SARA and association with SMAD4, allowing translocation to the nucleus. There, it binds to promoters and forms a transcription repressor complex. The protein can also be phosphorylated by activin type 1 receptor kinase to mediate activin signaling. Alternative splicing results in multiple transcript variants.