Learn More
ABclonal Technology Phospho-(Ser/Thr) ATM/ATR Substrate Rabbit pAb
SDP
Catalog No. 5000111985
Encompass_Preferred
The functionally related ATM (ataxia telangiectasia-mutated) and ATR (ATM-Rad3-related) protein kinases are critical regulators of DNA damage responses in mammalian cells. ATM and ATR share highly overlapping substrate specificities and show a strong preference for the phosphorylation of Serine (S) or Threonine (T) residues followed by Gln. It also called SQ or TQ consensus sites.
Catalog No. 50-001-11985 Supplier ABclonal Technology Supplier No. AP09331000UL
The functionally related ATM (ataxia telangiectasia-mutated) and ATR (ATM-Rad3-related) protein kinases are critical regulators of DNA damage responses in mammalian cells. ATM and ATR share highly overlapping substrate specificities and show a strong preference for the phosphorylation of Serine (S) or Threonine (T) residues followed by Gln. It also called SQ or TQ consensus sites.