Learn More
ABclonal Technology Phospho-SMO-S616 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP093820UL
Encompass_Preferred
The protein encoded by this gene is a G protein-coupled receptor that interacts with the patched protein, a receptor for hedgehog proteins. The encoded protein tranduces signals to other proteins after activation by a hedgehog protein/patched protein complex.
Catalog No. 50-001-11845