Learn More
ABclonal Technology Phospho-AURKA-T288/AURKB-T232/AURKC-T198 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP0948500UL
Encompass_Preferred
The protein encoded by this gene is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression. A processed pseudogene of this gene has been found on chromosome 1, and an unprocessed pseudogene has been found on chromosome 10. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq, Jul 2008]
Catalog No. 50-001-11367