Learn More
MSC Aqua-Pure Water Filter Cartridge, 5 micron, Polypropylene, 2/Pk.
Catalog No. 501113513
Encompass
This Aqua-Pure water filter cartridge is designed to effectively reduce dirt and rust in water. Featuring a 5 micron rating and made from polypropylene, it ensures clean water filtration for various applications.
  • Reduces dirt and rust
  • Features a graded-density design with proprietary polypropylene manufacturing
  • Grooved surface increases total surface area for longer cartridge life
  • Rigid construction helps eliminate sediment unloading due to water pressure variances
  • System materials are FDA CFR-21 compliant
  • Measures 9.75 in long with a 2.5 in outside diameter
  • Operates between 40°F and 100°F with a maximum flow rate of 8.00 GPM
  • Provides an average cartridge life of 6 months
Catalog No. 50-111-3513 Supplier MSC Supplier No. 07426356
This Aqua-Pure water filter cartridge is designed to effectively reduce dirt and rust in water. Featuring a 5 micron rating and made from polypropylene, it ensures clean water filtration for various applications.
  • Reduces dirt and rust
  • Features a graded-density design with proprietary polypropylene manufacturing
  • Grooved surface increases total surface area for longer cartridge life
  • Rigid construction helps eliminate sediment unloading due to water pressure variances
  • System materials are FDA CFR-21 compliant
  • Measures 9.75 in long with a 2.5 in outside diameter
  • Operates between 40°F and 100°F with a maximum flow rate of 8.00 GPM
  • Provides an average cartridge life of 6 months