Learn More
ABclonal Technology Phospho-STING/TMEM173-S366 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP12231000UL
Encompass_Preferred
This gene encodes a five transmembrane protein that functions as a major regulator of the innate immune response to viral and bacterial infections. The encoded protein is a pattern recognition receptor that detects cytosolic nucleic acids and transmits signals that activate type I interferon responses. The encoded protein has also been shown to play a role in apoptotic signaling by associating with type II major histocompatibility complex. Mutations in this gene are the cause of infantile-onset STING-associated vasculopathy. Alternate splicing results in multiple transcript variants.
Catalog No. 50-001-12100