Learn More
ABclonal Technology Phospho-IRAK4-Thr345/Ser346 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP1351500UL
Encompass_Preferred
This gene encodes a kinase that activates NF-kappaB in both the Toll-like receptor (TLR) and T-cell receptor (TCR) signaling pathways. The protein is essential for most innate immune responses. Mutations in this gene result in IRAK4 deficiency and recurrent invasive pneumococcal disease. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-11786