Learn More
ABclonal Technology Phospho-TIFA-T9 Rabbit pAb
SDP
Catalog No. 5000112015
Encompass_Preferred
This gene encodes an adapter protein involved in adaptive and innate immunity. This protein includes a forkhead-associated (FHA) domain that specifically binds to phosphorylated serine and threonine residues. In response to bacterial infection, the encoded host cell protein undergoes an intermolecular interaction between the FHA domain and a phosphorylated threonine that leads to protein oligomerization and stimulation of the NF-kappa B and other downstream signaling pathways. This protein exhibits reduced expression in hepatocellular carcinoma and may suppress hepatocellular carcinoma progression. This protein may also play a role in the DNA damage response.
Catalog No. 50-001-12015 Supplier ABclonal Technology Supplier No. AP13951000UL
This gene encodes an adapter protein involved in adaptive and innate immunity. This protein includes a forkhead-associated (FHA) domain that specifically binds to phosphorylated serine and threonine residues. In response to bacterial infection, the encoded host cell protein undergoes an intermolecular interaction between the FHA domain and a phosphorylated threonine that leads to protein oligomerization and stimulation of the NF-kappa B and other downstream signaling pathways. This protein exhibits reduced expression in hepatocellular carcinoma and may suppress hepatocellular carcinoma progression. This protein may also play a role in the DNA damage response.