Learn More
ABclonal Technology Phospho-p90Rsk/RSK1/RPS6KA1-T348 Rabbit pAb
Supplier: ABclonal Technology AP1397500UL
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.