Learn More
ABclonal Technology Phospho-AKT2 specific-S474 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP140320UL
Encompass_Preferred
This gene is a putative oncogene encoding a protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains, which is involved in signaling pathways. The gene serves as an oncogene in the tumorigenesis of cancer cells For example, its overexpression contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. The encoded protein is a general protein kinase capable of phophorylating several known proteins, and has also been implicated in insulin signaling.
Catalog No. 50-001-11199