Learn More
ABclonal Technology Phospho-Acetyl CoA Carboxylase-S79 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP1410500UL
Encompass_Preferred
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5 sequence and encoding distinct isoforms have been found for this gene.
Catalog No. 50-001-11994