Learn More
ABclonal Technology Phospho-BRD4-T249 Rabbit pAb
SDP
Catalog No. 5000112405
Encompass_Preferred
The protein encoded by this gene is homologous to the murine protein MCAP, which associates with chromosomes during mitosis, and to the human RING3 protein, a serine/threonine kinase. Each of these proteins contains two bromodomains, a conserved sequence motif which may be involved in chromatin targeting. This gene has been implicated as the chromosome 19 target of translocation t(15,19)(q13,p13.1), which defines an upper respiratory tract carcinoma in young people. Two alternatively spliced transcript variants have been described.
Catalog No. 50-001-12405 Supplier ABclonal Technology Supplier No. AP15151000UL
The protein encoded by this gene is homologous to the murine protein MCAP, which associates with chromosomes during mitosis, and to the human RING3 protein, a serine/threonine kinase. Each of these proteins contains two bromodomains, a conserved sequence motif which may be involved in chromatin targeting. This gene has been implicated as the chromosome 19 target of translocation t(15,19)(q13,p13.1), which defines an upper respiratory tract carcinoma in young people. Two alternatively spliced transcript variants have been described.