Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AP1867-2-carboxymeth 100mg | 2230613-03-1 | 100 MG
SDP

Catalog No. 5000227445
Encompass_Preferred

AP1867-2-(carboxymethoxy) is a PROTAC FKBP12-binding moiety based on the synthetic FKBP12F36V-directed ligand AP1867, intended for research use in assembling dTAG molecules that engage FKBP12F36V and CRBN in vitro.

  • High purity for reliable assay results.
  • Molecular weight 693.78 g/mol.
  • Chemical formula C38H47NO11.
  • Designed to link to CRBN ligands to form dTAG molecules.
  • Available in multiple pack sizes for assay development.

Catalog No. 50-002-27445 Supplier Medchemexpress LLC Supplier No. HY114420100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AP1867-2-(carboxymethoxy) is a PROTAC FKBP12-binding moiety based on the synthetic FKBP12F36V-directed ligand AP1867, intended for research use in assembling dTAG molecules that engage FKBP12F36V and CRBN in vitro.

  • High purity for reliable assay results.
  • Molecular weight 693.78 g/mol.
  • Chemical formula C38H47NO11.
  • Designed to link to CRBN ligands to form dTAG molecules.
  • Available in multiple pack sizes for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.